missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLK2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TLK2.
Brand: Abnova PAB3319.400uL
This item is not returnable.
View return policy
Description
The Tousled-like kinases, first described in Arabidopsis, are nuclear serine/threonine kinases that are potentially involved in the regulation of chromatin assembly.[supplied by OMIM
Specifications
| TLK2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TLK2. | |
| In PBS (0.09% sodium azide) | |
| MGC44450/PKU-ALPHA | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 11011 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| TLK2 | |
| TLK2 | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human TLK2. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction