Learn More
TMEM147, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TMEM147.
Brand:  Abnova PAB25760.100ug
Product Code. 16055937
  • € 619.00 / 100µg
Specifications
TMEM147
Polyclonal
Unconjugated
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
TMEM147
TMEM147
A synthetic peptide corresponding to 13 amino acids at internal region of human TMEM147.
100 μg
Primary
Human, Mouse, Rat
Liquid
ELISA, Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of TMEM147.
In PBS (0.02% sodium azide)
MGC1936/NIFIE14
Rabbit
Peptide affinity purification
RUO
10430
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.