missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TMEM5.
Brand: Abnova PAB16311.50ug
This item is not returnable.
View return policy
Specifications
| TMEM5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TMEM5. | |
| In PBS (0.09% sodium azide) | |
| HP10481 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 10329 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| TMEM5 | |
| TMEM5 | |
| A synthetic peptide (conjugated with KLH) corresponding to human TMEM5. | |
| 50 μg | |
| Primary | |
| Extracellular domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction