missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM59L Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TMEM59L.
Brand: Abnova PAB25711.100ug
This item is not returnable.
View return policy
Description
This gene encodes a predicted type-I membrane glycoprotein. The encoded protein may play a role in functioning of the central nervous system. [provided by RefSeq
Specifications
| TMEM59L | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| TMEM59L | |
| TMEM59L | |
| A synthetic peptide corresponding to 19 amino acids at C-terminus of human TMEM59L. | |
| 100 μg | |
| Primary | |
| Multiple isoforms of TMEM59L are known to exist. TMEM59L antibody is predicted to not cross-react TMEM59. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of TMEM59L. | |
| In PBS (0.02% sodium azide) | |
| BSMAP/C19orf4 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 25789 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction