Learn More
TNFRSF10B Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF10B.
Brand: Abnova PAB12830.100ug
Description
The protein encoded by this gene is a member of the TNF-receptor superfamily, and contains an intracellular death domain. This receptor can be activated by tumor necrosis factor-related apoptosis inducing ligand (TNFSF10/TRAIL/APO-2L), and transduces an apoptosis signal. Studies with FADD-deficient mice suggested that FADD, a death domain containing adaptor protein, is required for the apoptosis mediated by this protein. Two transcript variants encoding different isoforms and one noncoding transcript have been found for this gene. [provided by RefSeq
Specifications
| TNFRSF10B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF10B. | |
| In PBS (0.02% sodium azide) | |
| CD262/DR5/KILLER/KILLER/DR5/TRAIL-R2/TRAILR2/TRICK2/TRICK2A/TRICK2B/TRICKB/ZTNFR9 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody has no cross reaction to DR4. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| TNFRSF10B | |
| TNFRSF10B | |
| A synthetic peptide corresponding to C-terminus of human TNFRSF10B. | |
| RUO | |
| 8795 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only