Learn More
TNFRSF10D Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF10D.
Brand: Abnova PAB12831.100ug
Description
The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor contains an extracellular TRAIL-binding domain, a transmembrane domain, and a truncated cytoplamic death domain. This receptor does not induce apoptosis, and has been shown to play an inhibitory role in TRAIL-induced cell apoptosis. [provided by RefSeq
Specifications
| TNFRSF10D | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF10D. | |
| In PBS (0.02% sodium azide) | |
| CD264/DCR2/TRAILR4/TRUNDD | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| TNFRSF10D | |
| TNFRSF10D | |
| A synthetic peptide corresponding to amino acids 249-263 of human TNFRSF10D. | |
| RUO | |
| 8793 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only