Learn More
TNFSF13 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TNFSF13.
Brand: Abnova PAB13084.100ug
Description
The protein encoded by this gene is a member of the tumor necrosis factor (TNF) ligand family. This protein is a ligand for TNFRSF17/BCMA, a member of the TNF receptor family. This protein and its receptor are both found to be important for B cell development. In vitro experiments suggested that this protein may be able to induce apoptosis through its interaction with other TNF receptor family proteins such as TNFRSF6/FAS and TNFRSF14/HVEM. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. Other transcripts that skip the last exon of the upstream gene (TNFSF12) and continue with the second exon of this gene have been identified; such read-through transcripts are contained in GeneID 407977, TNFSF12-TNFSF13. [provided by RefSeq
Specifications
| TNFSF13 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TNFSF13. | |
| In PBS (0.02% sodium azide) | |
| APRIL/CD256/TALL2/TRDL-1/UNQ383/PRO715/ligand | |
| Rabbit | |
| 100 μg | |
| Primary | |
| A 27 KDa band can be detected. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (4-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| TNFSF13 | |
| TNFSF13 | |
| A synthetic peptide corresponding to internal region of human TNFSF13. | |
| RUO | |
| 8741 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only