missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNPO3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TNPO3.
Brand: Abnova PAB23434.100uL
This item is not returnable.
View return policy
Description
TNPO3 is a nuclear import receptor for serine/arginine-rich (SR) proteins, which are essential precursor-mRNA splicing factors (Kataoka et al., 1999 [PubMed 10366588]).[supplied by OMIM
Sequence: VRCVGKGSAALLQPLVTQMVNVYHVHQHSCFLYLGSILVDEYGMEEGCRQGLLDMLQALCIPTFQLLEQQNGLQNHPDTVDDLFRLATRFIQRSPVTLLRSQVVIPILQWAIASTTLDHSpecifications
| TNPO3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TNPO3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| IPO12/MTR10A/TRN-SR/TRN-SR2/TRNSR | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23534 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| TNPO3 | |
| TNPO3 | |
| Recombinant protein corresponding to amino acids of human TNPO3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction