Learn More
TNRC4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TNRC4.
Brand: Abnova PAB21916.100uL
Description
Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif (RRM) domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. While several transcript variants may exist for this gene, the full-length nature of only one has been biologically validated to date. [provided by RefSeq
Sequence: PVSAIPAALGVNGYSPVPTQPTGQPAPDALYPNGVHPYPAQSPAAPVDPLQQAYAGMQHYTAAYPAAYSLVAPAFPQPPALVAQQPPPPPQQQQQQQQQQQQQQQREGPDGCNIFIYHLPQEFTDSEILQMFVSpecifications
| TNRC4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TNRC4. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| BRUNOL1/CAGH4/CELF3/ERDA4/MGC57297 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 11189 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| TNRC4 | |
| TNRC4 | |
| Recombinant protein corresponding to amino acids of human TNRC4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only