Learn More
TOB1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human TOB1.
Brand: Abnova PAB29253.100uL
Description
This gene encodes a member of the tob/btg1 family of anti-proliferative proteins that have the potential to regulate cell growth. When exogenously expressed, this protein supresses cell growth in tissue culture. The protein undergoes phophorylation by a serine/threonine kinase, 90kDa ribosomal S6 kinase. Interactions of this protein with the v-erb-b2 erythroblastic leukemia viral oncogene homolog 2 gene product p185 interferes with growth suppression. This protein inhibits T cell proliferation and transcription of cytokines and cyclins. The protein interacts with both mothers against decapentaplegic Drosophila homolog 2 and 4 to enhance their DNA binding activity. This interaction inhibits interleukin 2 transcription in T cells. [provided by RefSeq
Specifications
| TOB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human TOB1. | |
| In PBS, pH 7.4 (without Mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| APRO6/MGC104792/MGC34446/PIG49/TOB/TROB/TROB1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 10140 | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user. | |
| TOB1 | |
| TOB1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S164 of human TOB1. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of TOB1 only when phosphorylated at serine 164. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only