missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOC1 Rabbit anti-Thale cress, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TOC1.
Brand: Abnova PAB16033.100ug
This item is not returnable.
View return policy
Specifications
| TOC1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TOC1. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| APRR1/MFB13.13/MFB13_13/PRR1/PSEUDO-RESPONSE REGULATOR 1/TIMING OF CAB EXPRESSION 1/TIMING OF CAB1 1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Thale cress | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| TOC1 | |
| TOC1 | |
| A synthetic peptide corresponding to N-terminus of Arabidopsis thaliana TOC1. | |
| RUO | |
| 836259 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction