Learn More
TOM1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TOM1
Brand: Abnova PAB28480.100uL
Description
This gene was identified as a target of the v-myb oncogene. The encoded protein shares its N-terminal domain in common with proteins associated with vesicular trafficking at the endosome. It is recruited to the endosomes by its interaction with endofin. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: LSGDTPIAPTPEQIGKLRSELEMVSGNVRVMSEMLTELVPTQAEPADLELLQELNRTCRAMQQRVLELIPQIANEQLTEELLIVNDNLNNVFLRHERFERFRTGQTTKAPSEAEPAADLIDMGPDPAATGNLSSQLAGMNLGSSSVRAGLQSLEASGRLEDEFDMFALTRGSSLADQSpecifications
| TOM1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TOM1 | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| FLJ33404 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10043 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot, Western Blot | |
| Unconjugated | |
| Immunohistochemistry(1:50-1:200) Western Blot(1:250-1:500) Immunofluorescence(1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| TOM1 | |
| TOM1 | |
| Recombinant protein corresponding to amino acids of human TOM1 | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only