missing translation for 'onlineSavingsMsg'
Learn More

TOMM70A Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Product Code. 16089596
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16089596 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16089596 Supplier Abnova Supplier No. PAB11995.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of TOMM70A.

The translocase of outer mitochondrial membrane (TOM) complex is a multisubunit complex involved in the recognition, unfolding, and translocation of preproteins into the mitochondria. See TIM17A (MIM 605057).[supplied by OMIM

Specifications

Antigen TOMM70A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TOMM70A.
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene TOMM70A
Gene Alias FLJ90470
Gene Symbols TOMM70A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-100 of human TOMM70A.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9868
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.