missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53BP1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant TP53BP1.
Brand: Abnova PAB12507.100uL
This item is not returnable.
View return policy
Specifications
| TP53BP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant TP53BP1. | |
| In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide) | |
| 53BP1/FLJ41424/MGC138366/p202 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody is specific to human 53BP1 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Immunocytochemistry (1:200-1:1000) Immunofluorescence (1:50-1:200) Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user. | |
| TP53BP1 | |
| TP53BP1 | |
| Recombinant protein corresponding to human TP53BP1. | |
| RUO | |
| 7158 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction