missing translation for 'onlineSavingsMsg'
Learn More

TRAF4, Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16018686
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16018686 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16018686 Supplier Abnova Supplier No. PAB0277.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of TRAF4.

This gene encodes a member of the TNF receptor associated factor (TRAF) family. TRAF proteins are associated with, and mediate signal transduction from members of the TNF receptor superfamily. The encoded protein has been shown to interact with neurotrophin receptor, p75 (NTR/NTSR1), and negatively regulate NTR induced cell death and NF-kappa B activation. This protein has been found to bind to p47phox, a cytosolic regulatory factor included in a multi-protein complex known as NAD(P)H oxidase. This protein thus, is thought to be involved in the oxidative activation of MAPK8/JNK. Alternatively spliced transcript variants have been observed but the full-length nature of only one has been determined. [provided by RefSeq

Sequence: MPGFDYKFLEKPKRRLL

Specifications

Antigen TRAF4
Applications Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TRAF4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene TRAF4
Gene Alias CART1/MLN62/RNF83
Gene Symbols TRAF4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-17 of human TRAF4.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9618
Target Species Bovine, Human, Mouse, Rat, Zebrafish
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.