Learn More
TRIM25 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TRIM25.
Brand: Abnova PAB24887.100uL
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the cytoplasm. The presence of potential DNA-binding and dimerization-transactivation domains suggests that this protein may act as a transcription factor, similar to several other members of the TRIM family. Expression of the gene is upregulated in response to estrogen, and it is thought to mediate estrogen actions in breast cancer as a primary response gene. [provided by RefSeq
Specifications
| TRIM25 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| TRIM25 | |
| TRIM25 | |
| A synthetic peptide corresponding to TRIM25. | |
| 100 μL | |
| Primary | |
| TRIM25 polyclonal antibody detects endogenous levels of TRIM25 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of TRIM25. | |
| In 1X PBS, pH 7.2 (0.05% sodium azide) | |
| EFP/RNF147/Z147/ZNF147 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 7706 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only