Learn More
TRPA1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TRPA1.
Brand: Abnova PAB27810.50ug
Description
The structure of the protein encoded by this gene is highly related to both the protein ankyrin and transmembrane proteins. The specific function of this protein has not yet been determined; however, studies indicate the function may involve a role in signal transduction and growth control. [provided by RefSeq
Specifications
| TRPA1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TRPA1. | |
| In PBS (0.09% sodium azide) | |
| ANKTM1 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 8989 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (7.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| TRPA1 | |
| TRPA1 | |
| A synthetic peptide corresponding to 19 amino acid at internal region of human TRPA1. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only