Learn More
TSG101 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TSG101.
Brand: Abnova PAB1759.400uL
Description
The protein encoded by this gene belongs to a group of apparently inactive homologs of ubiquitin-conjugating enzymes. The gene product contains a coiled-coil domain that interacts with stathmin, a cytosolic phosphoprotein implicated in tumorigenesis. The protein may play a role in cell growth and differentiation and act as a negative growth regulator. In vitro steady-state expression of this tumor susceptibility gene appears to be important for maintenance of genomic stability and cell cycle regulation. Mutations and alternative splicing in this gene occur in high frequency in breast cancer and suggest that defects occur during breast cancer tumorigenesis and/or progression. [provided by RefSeq
Specifications
| TSG101 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TSG101. | |
| In PBS (0.09% sodium azide) | |
| TSG10/VPS23 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 7251 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| TSG101 | |
| TSG101 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human TSG101. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only