Learn More
TSLP, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TSLP.
Brand: Abnova PAB13435.100ug
Description
This gene encodes a hemopoietic cytokine proposed to signal through a heterodimeric receptor complex composed of the thymic stromal lymphopoietin receptor and the IL-7R alpha chain. It mainly impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes and enhances the maturation of CD11c(+) dendritic cells. Alternative splicing of this gene results in two transcript variants. [provided by RefSeq]
Specifications
| TSLP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TSLP. | |
| In PBS (0.02% sodium azide) | |
| TSLP | |
| A synthetic peptide corresponding to internal region 19 amino acids of human TSLP. | |
| RUO | |
| 85480 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| TSLP | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only