Learn More
TTBK2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TTBK2.
Brand: Abnova PAB27186.100ug
Description
This gene encodes a serine-threonine kinase that putatively phosphorylates tau and tubulin proteins. Mutations in this gene cause spinocerebellar ataxia type 11 (SCA11); a neurodegenerative disease characterized by progressive ataxia and atrophy of the cerebellum and brainstem. [provided by RefSeq
Specifications
| TTBK2 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| TTBK2 | |
| TTBK2 | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human TTBK2. | |
| 100 μg | |
| Primary | |
| TTBK2 antibody is human specific. At least four isoforms of TTBK2 are known to exist; this antibody will detect only the two largest isoforms. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of TTBK2. | |
| In PBS (0.02% sodium azide) | |
| KIAA0847/SCA11/TTBK | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 146057 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only