missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBB, Rabbit, DyLight 549, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TUBB.
Brand: Abnova PAB15099.100uL
This item is not returnable.
View return policy
Specifications
| TUBB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TUBB. | |
| In 50mM sodium borate | |
| M40/MGC117247/MGC16435/OK/SW-cl.56/TUBB1/TUBB5 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This is specific to beta tubulin. This antibody can be used for Western Blot where a band at ∼50 KDa is seen. | |
| Store at 4°C. Do not freeze. |
| Western Blot | |
| DyLight 549 | |
| Western Blot (1:1000-1:10000) The optimal working dilution should be determined by the end user. | |
| TUBB | |
| TUBB | |
| A synthetic peptide corresponding to amino acids 1-100 of human TUBB. | |
| RUO | |
| 203068 | |
| Chicken, Human, Mouse, Pig, Rat, Xenopus species, Zebrafish | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction