missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBB2A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TUBB2A.
Brand: Abnova PAB0379.100ug
This item is not returnable.
View return policy
Description
Sequence: DLVSEYQQYQDATADEQGESpecifications
| TUBB2A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TUBB2A. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| TUBB/TUBB2/dJ40E16.7 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Bovine, Canine, Chicken, Frog, Human, Monkey, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| TUBB2A | |
| TUBB2A | |
| A synthetic peptide corresponding to amino acids 417-435 of human TUBB2A. | |
| RUO | |
| 7280 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction