missing translation for 'onlineSavingsMsg'
Learn More

TUBB3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Product Code. 16030698
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16030698 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16030698 Supplier Abnova Supplier No. PAB7877.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of TUBB3.

Specifications

Antigen TUBB3
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TUBB3.
Dilution ELISA (1:2000) Immunocytochemistry (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Gene TUBB3
Gene Alias MC1R/TUBB4/beta-4
Gene Symbols TUBB3
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to residues surrounding S444 of human TUBB3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10381
Test Specificity This sequence is not found in bI or bII-tubulin isotypes, but is well conserved in bIII-tubulins from rat and mouse.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.