missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tusc5 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Tusc5.
Brand: Abnova PAB15951.100ug
This item is not returnable.
View return policy
Specifications
| Tusc5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Tusc5. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| C130069F04Rik/LOST1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Tusc5 | |
| Tusc5 | |
| A synthetic peptide corresponding to N-terminus of mouse Tusc5. | |
| RUO | |
| 237858 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction