Learn More
TYSND1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TYSND1.
Brand: Abnova PAB22755.100uL
Description
All peroxisomal proteins are synthesized in the cytosol, and 2 distinct peroxisomal targeting signals (PTSs), the C-terminal PTS1 and N-terminal PTS2, are used for transport of these proteins into peroxisomes. Proteolytic cleavage of the N-terminal targeting sequence of PTS2 proteins accompanies import into peroxisomes, and many PTS1 proteins undergo C-terminal processing once in the peroxisomal matrix. TYSND1 processes both PTS1 and PTS2 proteins involved in beta-oxidation of fatty acids (Kurochkin et al., 2007 [PubMed 17255948]).[supplied by OMIM
Sequence: ATQETCPYDIAVVSLEEDLDDVPIPVPAEHFHEGEAVSVVGFGVFGQSCGPSVTSGILSAVVQVNGTPVMLQTTCAVSpecifications
| TYSND1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TYSND1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ40378/MGC131934/MGC34695 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 219743 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| TYSND1 | |
| TYSND1 | |
| Recombinant protein corresponding to amino acids of human TYSND1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only