Learn More
UBE2V2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of UBE2V2.
Brand: Abnova PAB0193.100ug
Description
Ubiquitin-conjugating enzyme E2 variant proteins constitute a distinct subfamily within the E2 protein family. They have sequence similarity to other ubiquitin-conjugating enzymes but lack the conserved cysteine residue that is critical for the catalytic activity of E2s. The protein encoded by this gene also shares homology with ubiquitin-conjugating enzyme E2 variant 1 and yeast MMS2 gene product. It may be involved in the differentiation of monocytes and enterocytes. [provided by RefSeq
Specifications
| UBE2V2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of UBE2V2. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| DDVIT1/DDVit-1/EDAF-1/EDPF-1/EDPF1/MMS2/UEV-2/UEV2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| UBE2V2 | |
| UBE2V2 | |
| A mixture of synthetic peptides corresponding to amino acids 12-28 of human UBE2V2. | |
| RUO | |
| 7336 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only