Learn More
USP25 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of USP25.
Brand: Abnova PAB4213.400uL
Description
Ubiquitin (MIM 191339) is a highly conserved 76-amino acid protein involved in regulation of intracellular protein breakdown, cell cycle regulation, and stress response. Ubiquitin is released from degraded proteins by disassembly of the polyubiquitin chains, which is mediated by ubiquitin-specific proteases (USPs), such as USP25 (Valero et al., 1999 [PubMed 10644437]).[supplied by OMIM
Specifications
| USP25 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of USP25. | |
| In PBS (0.09% sodium azide) | |
| USP21 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 29761 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
| USP25 | |
| USP25 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human USP25. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only