Learn More
VEGFC Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of VEGFC.
Brand:  Abnova PAB3867.400uL
Product Code. 16098827
  • € 388.00 / 400µL
Description
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D. [provided by RefSeq
Specifications
Specifications
VEGFC
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of VEGFC.
In PBS (0.09% sodium azide)
Flt4-L/VRP
Rabbit
Protein A purification
RUO
7424
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry, Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
VEGFC
VEGFC
A synthetic peptide (conjugated with KLH) corresponding to internal region of human VEGFC.
400 μL