Learn More
VEGFC Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of VEGFC.
Brand: Abnova PAB3867.400uL
Description
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D. [provided by RefSeq
Specifications
| VEGFC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of VEGFC. | |
| In PBS (0.09% sodium azide) | |
| Flt4-L/VRP | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 7424 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| VEGFC | |
| VEGFC | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human VEGFC. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |