Learn More
VN1R1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of VN1R1.
Brand: Abnova PAB27714.50ug
Description
Pheromones are chemical signals that elicit specific behavioral responses and physiologic alterations in recipients of the same species. The protein encoded by this gene is similar to pheromone receptors and is primarily localized to the olfactory mucosa. An alternate splice variant of this gene is thought to exist, but its full length nature has not been determined. [provided by RefSeq
Specifications
| VN1R1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of VN1R1. | |
| In PBS (0.09% sodium azide) | |
| V1RL1/VNR19I1/ZVNH1/ZVNR1 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 57191 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (8 ug/mL) The optimal working dilution should be determined by the end user. | |
| VN1R1 | |
| VN1R1 | |
| A synthetic peptide corresponding to 18 amino acid at cytoplasmic domain of human VN1R1. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only