Learn More
WAC Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of WAC.
Brand: Abnova PAB27413.100ug
Description
The protein encoded by this gene contains a WW domain, which is a protein module found in a wide range of signaling proteins. This domain mediates protein-protein interactions and binds proteins containing short linear peptide motifs that are proline-rich or contain at least one proline. This gene product shares 94% sequence identity with the WAC protein in mouse, however, its exact function is not known. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Specifications
| WAC | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) Immunofluorescence (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| WAC | |
| WAC | |
| A synthetic peptide corresponding to 19 amino acids at internal region of human WAC. | |
| 100 μg | |
| Primary | |
| At least two isoforms of WAC are known to exist; this antibody will detect the larger isoform. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of WAC. | |
| In PBS (0.02% sodium azide) | |
| BM-016/MGC10753/PRO1741/Wwp4/bA48B24/bA48B24.1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 51322 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only