Learn More
WFDC1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant WFDC1.
Brand: Abnova PAB22551.100uL
Description
This gene encodes a member of the WAP-type four disulfide core domain family. The WAP-type four-disulfide core domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. The encoded protein shares 81% amino acid identity with the rat ps20 protein, which was originally identified as a secreted growth inhibitor. This gene is mapped to chromosome 16q24, an area of frequent loss of heterozygosity in cancers, including prostate, breast and hepatocellular cancers and Wilms' tumor. Owing to its location and a possible growth inhibitory property of its gene product, this gene is suggested to be a tumor suppressor gene. [provided by RefSeq]
Sequence: LLCPSGYECHILSPGDVAEGIPNRGQCVKQRRQADGRILRHKLYKEYPEGDSKNVAEPGRGQQKHFQSpecifications
| WFDC1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant WFDC1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| PS20 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 58189 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| WFDC1 | |
| WFDC1 | |
| Recombinant protein corresponding to amino acids of human WFDC1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only