missing translation for 'onlineSavingsMsg'
Learn More

WHSC1L1 Rabbit anti-Bovine, Human, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16079586
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16079586 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16079586 Supplier Abnova Supplier No. PAB11983.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of WHSC1L1.

This gene is related to the Wolf-Hirschhorn syndrome candidate-1 gene and encodes a protein with PWWP (proline-tryptophan-tryptophan-proline) domains. The function of the protein has not been determined. Two alternatively spliced variants have been described. [provided by RefSeq

Specifications

Antigen WHSC1L1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of WHSC1L1.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Gene WHSC1L1
Gene Alias DKFZp667H044/FLJ20353/MGC126766/MGC142029/NSD3/pp14328
Gene Symbols WHSC1L1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 300-400 of human WHSC1L1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54904
Test Specificity This antibody recognize a band that corresponds to isoform 1 of NSD3.
Target Species Bovine, Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.