missing translation for 'onlineSavingsMsg'
Learn More

WNK4, Rabbit, Biotin, Polyclonal Antibody, Abnova™

Product Code. 16051417
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16051417 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16051417 Supplier Abnova Supplier No. PAB15075.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of WNK4.

This gene encodes a member of the WNK family of serine-threonine protein kinases. The kinase is part of the tight junction complex in kidney cells, and regulates the balance between NaCl reabsorption and K(+) secretion. The kinase regulates the activities of several types of ion channels, cotransporters, and exchangers involved in electrolyte flux in epithelial cells. Mutations in this gene result in pseudohypoaldosteronism type IIB

Specifications

Antigen WNK4
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against synthetic peptide of WNK4.
Dilution Western Blot (2 ug/ mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene WNK4
Gene Alias PHA2B/PRKWNK4
Gene Symbols WNK4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 200-300 of human WNK4.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65266
Test Specificity This antibody can be used to detect mouse and human (very weakly) WNK4 protein. NB600-284 is specific to WNK4 protein. This antibody is useful for Western blot where a band is seen at ∼135-155 kD. No other applications have been tested.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.