Learn More
WNT6 Rabbit anti-Bovine, Dog, Hamster, Horse, Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyconal antibody raised against synthetic peptide of WNT6.
Brand: Abnova PAB28362.50ug
Description
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is overexpressed in cervical cancer cell line and strongly coexpressed with another family member, WNT10A, in colorectal cancer cell line. The gene overexpression may play key roles in carcinogenesis. This gene and the WNT10A gene are clustered in the chromosome 2q35 region. The protein encoded by this gene is 97% identical to the mouse Wnt6 protein at the amino acid level. [provided by RefSeq
Specifications
| WNT6 | |
| Polyclonal | |
| Rabbit polyconal antibody raised against synthetic peptide of WNT6. | |
| In PBS (0.09% sodium azide) | |
| WNT6 | |
| A synthetic peptide corresponding to 20 amino acids at N-terminus of human WNT6. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/ml) The optimal working dilution should be determined by the end user. | |
| WNT6 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 7475 | |
| Bovine, Canine, Equine, Hamster, Human, Mouse, Rabbit, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only