missing translation for 'onlineSavingsMsg'
Learn More

Rabbit Serum Antibody, Novus Biologicals™

Product Code. 18108997 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18108997 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18108997 Supplier Novus Biologicals Supplier No. NBP244099

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Specifically detects Rabbit Serum in Rabbit samples, and it is validated for ELISA, Immunoprecipitation, Western Blotting

Rabbit Serum Polyclonal specifically detects Rabbit Serum in Rabbit samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit Serum
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Alias Rabbit Sera
Host Species Goat
Immunogen Rabbit Serum Antibody was produced by repeated immunizations with Rabbit serum proteins
Purification Method Delipidation and Defibrination
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute with 2.0 ml deionized water (or equivalent)
Target Species Rabbit
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.