missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit TNF-alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5670
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Rabbit samples. It is validated for Neutralization.
Specifications
| TNF-alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7124 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization | |
| 610331 | |
| Neutralization 0.4-2.4 ug/mL | |
| NP_001075732 | |
| TNF | |
| E. coli-derived recombinant rabbit TNF-alpha Leu79-Leu234 Accession # NP_001075732 | |
| 500 μg | |
| Primary | |
| Detects rabbit TNF-alpha in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant TNF-alpha from cotton rat, cat, cow, dog, guinea pig, horse, human, mouse, pig, rat, or rhesus is observed. No cross-reactivity with recombinant human (rh) 4-1BB Ligand, rhAPRIL, rhBAFF, rhEDA, rhEDA-A2, rhFas Ligand, rhGITR Ligand, rhLIGHT, rhLymphotoxin alpha 1 beta 2, rhLymphotoxin alpha 2 beta 1, rhOX40 Ligand, rhTRAIL, rhTRANCE, rhTWEAK, rhVEGI, recombinant mouse (rm) 4-1BB Ligand, rmBAFF, rmFas Ligand, rmGITR Ligand, rmLIGHT, rmOX40 Ligand, rmTRAIL, rmTRAIL-2L, rmTRANCE, or rmTWEAK is observed. | |
| Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction