missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABEP1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-47285B
This item is not returnable.
View return policy
Description
RABEP1 Polyclonal antibody specifically detects RABEP1 in Human samples. It is validated for Western Blot
Specifications
| RABEP1 | |
| Polyclonal | |
| Western Blot | |
| neurocrescin, rab GTPase-binding effector protein 1, RAB5EPRABPT5A, rabaptin, RAB GTPase binding effector protein 1, rabaptin-4, rabaptin-5, Rabaptin-5alpha, RABPT5rabaptin-5alpha, Renal carcinoma antigen NY-REN-17 | |
| The immunogen for this product maps to a region between residue 300 and 350 of human Rabaptin, RAB GTPase Binding Effector Protein 1 using the numbering given in entry NP_004694.2 (GeneID 9135). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9135 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction