Learn More
Invitrogen™ RABEP1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5118232
Product Code. 17431893
  • € 485.00 / 100µL
Description
RABEP1 Polyclonal Antibody for Western Blot, ICC/IF, IP
RABEP1 gene ontology annotations related to this gene include endocytosis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RABEP1
Polyclonal
PBS with 0.03% ProClin 300; pH 7
Q15276
RABEP1
E. coli-derived Human RABEP1 fragment.
100 μL
Primary
Human
Antibody
IgG
Immunoprecipitation, Western Blot, Immunocytochemistry
Unconjugated
RABEP1
neurocrescin; rab GTPase-binding effector protein 1; RAB5 effector protein; RAB5EP; rabaptin 5; rabaptin, RAB GTPase binding effector protein 1; rabaptin-4; Rabaptin-5; rabaptin-5alpha; Rabep1; RABPT5; Rabpt5a; Renal carcinoma antigen NY-REN-17
Rabbit
Antigen affinity chromatography, Protein A
RUO
9135
-20°C, Avoid Freeze/Thaw Cycles