missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabex5 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97267APC
This item is not returnable.
View return policy
Description
Rabex5 Polyclonal antibody specifically detects Rabex5 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Rabex5 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| FLJ32302, RAB guanine nucleotide exchange factor (GEF) 1, rab5 GDP/GTP exchange factor, Rabaptin-5-associated exchange factor for Rab5, Rabex-5, RABEX5rabex-5, RAP1 | |
| Produced in rabbits immunized with E. coli-derived Human Rabex5 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 27342 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction