missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABGAP1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44284AF350
This item is not returnable.
View return policy
Description
RABGAP1 Polyclonal antibody specifically detects RABGAP1 in Human samples. It is validated for Western Blot
Specifications
| RABGAP1 | |
| Polyclonal | |
| Western Blot | |
| DKFZp586D2123, GAP and centrosome-associated protein, GAPCENA, RAB GTPase activating protein 1, rab GTPase-activating protein 1, rab6 GTPase activating protein (GAP and centrosome-associated), Rab6 GTPase activating protein, GAPCenA, Rab6 GTPase-activating protein GAPCenA, RP11-123N4.2, TBC1 domain family, member 11, TBC1D11 | |
| The epitope recognized by this antibody maps to a region between residue 1 to 50 of human Rab GTPase-activating protein 1 using the numbering given in entry Q9Y3P9.3 (GeneID 23637). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23637 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction