missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RABGAP1 Polyclonal Antibody

Description
Immunogen sequence: EKQQTANKVEI EKIRQKVDDC ERCREFFNKE GRVKGISSTK EVLDEDTDEE KETLKN Highest antigen sequence identity to the following orthologs - mouse 91%, rat 89%.
RABGAP1 may act as a GTPase-activating protein of RAB6A and play a role in microtubule nucleation by centrosome. It may participate in a RAB6A-mediated pathway involved in the metaphase-anaphase transition.
Specifications
Specifications
| Antigen | RABGAP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RABGAP1 |
| Gene Accession No. | Q9Y3P9 |
| Gene Alias | G protein-coupled receptor 21; GAP and centrosome-associated protein; GAPCENA; Gpr21; HSPC094; Kiaa4104; mKIAA4104; RAB GTPase activating protein 1; rab GTPase-activating protein 1; rab6 GTPase activating protein (GAP and centrosome-associated); Rab6 GTPase activating protein, GAPCenA; rab6 GTPase-activating protein GAPCenA; RABGAP1; RP11-123N4.2; TBC1 domain family, member 11; TBC1D11 |
| Gene Symbols | RABGAP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?