Learn More
RABGAP1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14223-1-AP-150UL
 View more versions of this product
Product Code. 16822764
  • € 410.00 / 150µL
Description
The RABGAP1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RABGAP1. This antibody recognizes human antigen. The RABGAP1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RABGAP1
Polyclonal
Unconjugated
150 μL
23637
IgG
Western Blot, ELISA
233 μg/mL