missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABGGTA Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97779PE
This item is not returnable.
View return policy
Description
RABGGTA Polyclonal antibody specifically detects RABGGTA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RABGGTA | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| EC 2.5.1.60, Geranylgeranyl transferase type II subunit alpha, geranylgeranyl transferase type-2 subunit alpha, protein prenyltransferase alpha subunit repeat containing 3, PTAR3, Rab geranylgeranyltransferase subunit alpha, Rab geranyl-geranyltransferase subunit alpha, Rab geranylgeranyltransferase, alpha subunit, Rab GG transferase alpha, Rab GGTase alpha | |
| Produced in rabbits immunized with E. coli-derived Human RABGGTA fragment. | |
| 0.1 mL | |
| GPCR, Neuroscience, Signal Transduction | |
| 5875 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction