missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabies Virus Antibody (1C5cc), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11630IR-0.2ML
This item is not returnable.
View return policy
Description
Rabies Virus Monoclonal specifically detects Rabies Virus in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Rabies Virus | |
| Monoclonal | |
| DyLight 755 | |
| 50 mM Sodium Borate | |
| It has a virus-neutralizing activity in respect to the majority of standard RV and rabies-relative viruses: CVS, Lagos bat, Mokola, Flury lep, Duvenhage and the field isolates | |
| Virus | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1C5 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Mouse | |
| 0.2 mL | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction