missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabies Virus Antibody (Rab-50), Alexa Fluor™ 350 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63045AF350
This item is not returnable.
View return policy
Description
Rabies Virus Monoclonal antibody specifically detects Rabies Virus in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Rabies Virus | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Glycoprotein | |
| Purified virions of SAD-Vnukova strain followed by Pitman-Moore strain. | |
| 0.5 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, ELISA, Immunofluorescence | |
| Rab-50 | |
| 50mM Sodium Borate | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction