missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabies Virus Nucleoprotein Antibody (12) [Alexa Fluor™ 488] - (CVS-11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-33166AF488
This item is not returnable.
View return policy
Description
Rabies Virus Nucleoprotein Monoclonal antibody specifically detects Rabies Virus Nucleoprotein in Virus samples. It is validated for ELISA,Sandwich ELISA Detection
Specifications
| Rabies Virus Nucleoprotein | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Sandwich ELISA Detection | |
| 12 | |
| 50mM Sodium Borate | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rabies Virus Nucleoprotein (Uniprot#: Q8JXF6; Met1-Ser450). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction