missing translation for 'onlineSavingsMsg'
Learn More

RABL2A Antibody - BSA Free, Novus Biologicals™

Product Code. p-200062301 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18641272 0.02 mL 0.02mL
18609631 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18641272 Supplier Novus Biologicals Supplier No. NBP2943140.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Guinea Pig Polyclonal Antibody

RABL2A Polyclonal antibody specifically detects RABL2A in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABL2A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias FLJ78724, MGC117180, RAB, member of RAS oncogene family-like 2A, rab-like protein 2A
Host Species Guinea Pig
Immunogen Recombinant Protein of mouse RABL2A
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11159
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.