missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABL2A Antibody (OTI4A8), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73776AF532
This item is not returnable.
View return policy
Description
RABL2A Monoclonal specifically detects RABL2A in Human, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| RABL2A | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RABL2A | |
| Full length recombinant protein of human RABL2A (NP_009013) produced in HEK293T cells. | |
| Primary | |
| Human, Rat, Monkey, Primate | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI4A8 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| FLJ78724, MGC117180, RAB, member of RAS oncogene family-like 2A, rab-like protein 2A | |
| Mouse | |
| 0.1 mL | |
| 11159 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction