missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABL2A Antibody (OTI4A8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-03281
This item is not returnable.
View return policy
Description
RABL2A Monoclonal specifically detects RABL2A in Human, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| RABL2A | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100 | |
| RABL2A | |
| Full length human recombinant protein of human RABL2A (NP_009013) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Rat, Monkey, Primate | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI4A8 | |
| Unconjugated | |
| FLJ78724, MGC117180, RAB, member of RAS oncogene family-like 2A, rab-like protein 2A | |
| Mouse | |
| 25.9 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 11159 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction