missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABL2A Monoclonal Antibody (OTI1A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502021S
This item is not returnable.
View return policy
Description
RABL2A Monoclonal antibody specifically detects RABL2A in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| RABL2A | |
| Monoclonal | |
| 0.43 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UBK7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11159 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1A12 | |
| Unconjugated | |
| RABL2A | |
| 1110031N17Rik; AW549303; RAB, member of RAS oncogene family-like 2; RAB, member of RAS oncogene family-like 2A; RAB, member of RAS oncogene family-like 2B; RAB, member RAS oncogene family-like 2; Rabl2; RABL2A; Rabl2b; rab-like protein 2A; rab-like protein 2B; RP11-395L14.2 | |
| Full length human recombinant protein of human RABL2A produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction